Learn More
Medchemexpress LLC OSu-PEG4-VC-PAB-MMAE | 1mg
SDP
Supplier:  Medchemexpress LLC HY1484261MG
Encompass_Preferred
OSu-PEG4-VC-PAB-MMAE involves in GDP-FAmP4MMAE (Monomethylauristatin E) synthesis which is for GDP label via site-specific conjugation[1]
Catalog No. 50-002-01957