Learn More
Selleck Chemical LLC OSU-T315
SDP
Supplier:  Selleck Chemical LLC S68402MG
Encompass_Preferred
OSU-T315 (ILK-IN-2) is a potent Integrin-Linked Kinase (ILK) inhibitor with IC50 of 0 6 M OSU-T315 induces autophagy and apoptosis both of which are integral to its antiproliferative activity OSU-T315 exhibits anti-tumor activity
Catalog No. 50-000-02710