Learn More
Medchemexpress LLC Otenaproxesul | 1226895-20-0 | C21H19NO3S | 1 ML

Supplier: Medchemexpress LLC HY1502810MM/1ML
Otenaproxesul is a chemical compound identified by CAS number 1226895-20-0 and chemical formula C21H19NO3S. It is supplied as a light yellow to yellow solid with a high purity of 98.03%. This product is intended for laboratory use in research and manufacturing of substances, and it has not been validated for medical applications.
- Chemical name: 2-Naphthaleneacetic acid, 6-methoxy-α-methyl-, 4-(aminothioxomethyl)phenyl ester
- Molecular weight: 365.45
- Appearance: Light yellow to yellow solid
- Purity (HPLC): 98.03%
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage in solvent: -80°C for 2 years, -20°C for 1 year
- Identified uses: Laboratory chemicals, manufacture of substances
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.