Learn More
Medchemexpress LLC Otenaproxesul | 1226895-20-0 | C21H19NO3S | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15028100MG
Encompass_Preferred
Otenaproxesul (ATB-346) is an orally active non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and 2 (COX-1 and 2). It exhibits anti-inflammatory and antinociceptive activities.
  • Inhibits cyclooxygenase-1 and 2 (COX-1 and 2)
  • Exhibits anti-inflammatory properties
  • Shows antinociceptive activities
  • Reduces gastrointestinal toxicity
  • Inhibits human melanoma cell proliferation
  • Induces apoptosis of human melanoma cells
  • Inhibits melanoma tumor growth in vivo
  • Suppresses PGE2 levels and TXB2 synthesis
  • Reduces paw oedema
Catalog No. 50-003-75920