Learn More
Medchemexpress LLC Otenaproxesul | 1226895-20-0 | C21H19NO3S | 50 MG

Supplier: Medchemexpress LLC HY1502850MG
Otenaproxesul (ATB-346) is a laboratory chemical primarily intended for research use only. This solid substance is classified as hazardous, causing skin irritation, serious eye irritation, and potential respiratory tract irritation, requiring handling by qualified personnel in appropriate facilities.
- Research-grade laboratory chemical
- Solid appearance
- Stable under recommended storage conditions
- Causes skin irritation
- Causes serious eye irritation
- May cause respiratory irritation
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage for in solvent: -80°C for 2 years, -20°C for 1 year
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.