Learn More
Medchemexpress LLC Otenaproxesul | 1226895-20-0 | CB52627347 | C21H19NO3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY150285MG
Encompass_Preferred
Otenaproxesul is an orally active non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and 2 (COX-1 and 2). It possesses anti-inflammatory and antinociceptive activities, exhibiting properties similar to naproxen but with substantially reduced gastrointestinal toxicity.
  • Inhibits cyclooxygenase-1 and 2
  • Possesses anti-inflammatory and antinociceptive activities
  • Reduced gastrointestinal toxicity
  • Inhibits human melanoma cell proliferation
  • Induces apoptosis of human melanoma cells
  • Inhibits bone defect and histological characteristics related to periodontitis
Catalog No. 50-003-75583