Learn More
Medchemexpress LLC OTs-C6-OBn | 126519-80-0 | 98.1% | 250 MG

Supplier: Medchemexpress LLC 250MGHY130621
OTs-C6-OBn is an alkyl chain-based PROTAC linker. It is primarily used in the synthesis of PROTAC SGK3 degrader-1. PROTACs, or Proteolysis-targeting chimeras, are molecules that utilize the intracellular ubiquitin-proteasome system to selectively degrade target proteins. They function by connecting two different ligands: one for an E3 ubiquitin ligase and another for the target protein.
- Alkyl chain-based PROTAC linker
- Used in the synthesis of PROTAC SGK3 degrader-1
- Facilitates target protein degradation via the ubiquitin-proteasome system by linking an E3 ubiquitin ligase ligand to a target protein ligand
- Purity: 98.07%
- Appearance: Liquid, colorless to light yellow
- Molecular weight: 362.48
- Formula: C20H26O4S
- CAS number: 126519-80-0
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.