Learn More
Medchemexpress LLC HY-122181B 5mg Medchemexpress, OTS186935 (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY122181B5MG
Encompass_Preferred
Medchemexpress, HY-122181B 5mg OTS186935 (hydrochloride) CAS:0 OTS186935 hydrochloride is a potent protein methyltransferase SUV39H2 inhibitor with an IC50 of 6.49 nM. OTS186935 hydrochloride shows significant inhibition of tumor growth in mouse xenograft models without any detectable toxicity. OTS193320 hydrochloride regulates the production of γ-H2AX in cancer cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9920