Learn More
Selleck Chemical LLC OTS964-5mg
SDP
Supplier:  Selleck Chemical LLC S76485MG
Encompass_Preferred
OTS964 is a potent TOPK inhibitor with high affinity and selectivity and IC50 value is 28 nM. OTS964 is also a potent inhibitor of the cyclin-dependent kinase CDK11 with Kd of 40 nM. OTS964 treatment activates autophagy in glioma cells and induces apoptosis of human lung cancer cells in mouse xenografts.
Catalog No. 50-238-9760