Learn More
Cayman Chemical OTsP167hydrochlorIde 5mg
SDP
Supplier:  Cayman Chemical 168735 MG
Encompass_Preferred
A potent inhibitor of MELK (IC50 = 0.41 nM); blocks the phosphorylation of MELK-specific substrates and reduces the ability of MCF-7 breast cancer cells to invade and form spheroids in Matrigel; suppresses the growth of xenograft tumors in mice
Catalog No. 50-296-7427