Learn More
Selleck Chemical LLC OTSSP167 5mg 1431697-89-0 OTS167
SDP
Supplier:  Selleck Chemical LLC S71595MG
Encompass_Preferred
OTSSP167 (OTS167) is a highly potent MELK (maternal embryonic leucine zipper kinase) inhibitor with IC50 of 0.41 nM. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8033