Learn More
Waters Corp Cartridge Top Rack, 1 cc, 24-Position, for Otto SPEcialist
Supplier:  Waters Corp 186009716
Encompass_Preferred
This cartridge top rack is designed for use with the Otto SPEcialist system. It accommodates up to 24 1cc SPE cartridges, providing a convenient and organized solution for sample preparation workflows.
  • Fits up to 24 1cc SPE cartridges.
  • Designed for the Otto SPEcialist system.
  • Sits on top of the Otto Accessories stack.
Catalog No. 50-239-2955