Learn More
Apexbio Technology LLC OTX-015, 5mg. Cas: 202590-98-5 MFCD: MFCD26960949. BRD inhibitor
SDP
Supplier:  Apexbio Technology LLC A36925
Encompass_Preferred
OTX-015 is a potent inhibitor of BRD2, BRD3, and BRD4 with IC50 values range from 92 to 112 nM [1].BRD2, BRD3, and BRD4 belong to BET bromodomain family and play an important role in regulating transcription. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4826