Learn More
Medchemexpress LLC Oul35 (NSC39047) | 6336-34-1 | 99.8% | 256.26 g/mol | C14H12N2O3 | 10MG

Supplier: Medchemexpress LLC HY12351210MG
OUL35 (NSC39047) is a selective small-molecule inhibitor of ARTD10 (PARP10) used as a chemical probe in studies of ADP-ribosylation and DNA damage response. It inhibits ARTD10 with reported IC50 = 329 nM and is supplied as a high-purity solid for research applications.
- Selective ARTD10 (PARP10) inhibitor; IC50 329 nM.
- High purity: 99.8%.
- Molecular weight: 256.26 g/mol.
- Chemical formula: C14H12N2O3.
- Physical form: white to off-white solid.
- Solubility: soluble in DMSO for stock solutions.
- Suitable for cellular and biochemical assays probing PARP10 function.
- Available in research-scale quantities (mg scale).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.