Learn More
COLDER PRODUCTS COMPANY OUPLER BODY EA
Supplier:  COLDER PRODUCTS COMPANY PMCD170212
Encompass_Preferred
CPC Standard-Volume Quick-Disconnect Inline and Panel-Mount Hose-Barb Bodies For Use With 3 2mm EPDM Seal Overall Length 42 4 mm 1 67 in Almond Diameter Inner 0 125 in Includes Shut-off valves Hose Barb Valved In-line Coupling Body Height 0 89 in 22 6 mm Maximum Pressure 120 psi
Catalog No. 50-313-1404