Learn More
Optimize Technologies OPTI-MAX™ Outlet Cartridge Check Valve, 1/16" Ruby Ball/Sapphire Seat, SS Cartridge, HP/Agilent 1050, 1100, 1200 Low Flow Rate
SDP
Supplier:  Optimize Technologies 274701330
Encompass_Preferred
Opti-Max™ Cartridge Check Valves are designed to offer maximum performance and ease of use. From the sharp, consistent threads on OPTI-MAX™ custom housings, to the exacting standards used to match ball/seat sets in OPTI-MAX™ cartridges, every aspect of design and manufacturing reflects an unavoidable fact: when it comes to reliable check valve operation, quality and precision make a big difference. OPTI-MAX™ check valves offer many advantages over OEM check valve systems, and are better designed, better made and more reliable than any other after-market check valve product. Additionally, OPTI-MAX™ Free-Turn™ housings can be removed with the tubing still attached, accelerating check valve maintenance while reducing fitting wear.
Catalog No. 50-200-6631