Learn More
Invivogen OVA peptides, H-2Kb-restricted OVA MHC class I epitope
SDP
Supplier:  Invivogen VACSIN
Encompass
OVA 257-264 is a class I (Kb)-restricted peptide epitope of ovalbumin (OVA), presented by the class I major histocompatibility complex (MHC) molecule, H-2Kb [1]. It has been demonstrated that OVA 257-264 peptides can be used to detect a strong CD8+ cytolytic T cell response [2]. 
Catalog No. NC1070288