Learn More
Medchemexpress LLC OVA (329-337) | 276889-40-8 | 98.7% | 924.95 | 50 MG

Supplier: Medchemexpress LLC HYP253150MG
OVA (329-337) is a 9-aa core epitope (329-337) found in the C-terminal end of the OVA peptide. This product is intended for research use only. In vitro studies show that OVA (329-339) induces half-maximal proliferation with 25 μM in DO11.10 cells and 30 μM in OT-II cells.
- 9-aa core epitope (329-337) of the OVA peptide
- Purity of 98.73%
- Molecular weight of 924.95
- Chemical formula C38H60N12O15
- Appears as a solid, white to off-white color
- Sequence: Ala-Ala-His-Ala-Glu-Ile-Asn-Glu-Ala (AAHAEINEA)
- Ships at room temperature (continental US)
- Store powder at -80 °C for 2 years or -20 °C for 1 year
- Store in solvent at -80 °C for 6 months or -20 °C for 1 month (sealed, away from moisture)
- Soluble in H2O at 1.82 mg/mL (1.97 mM)
- Slightly soluble or insoluble in DMSO (< 1 mg/mL)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.