Learn More
BRINSEA PRODUCTS Ova-Easy 380 Advance Series II Cabinet Incubator
Supplier:  BRINSEA PRODUCTS MJ3823C
Encompass_Preferred
Like all 4 models in the Brinsea cabinet incubator range, the Ova-Easy 380 Advance Series II is designed to offer the best possible performance and is fitted with Brinsea’s proven Advance fully digital control system which is both accurate and easy to use. Incubation temperature (in °C or °F) and humidity are continuously displayed along with temperature alarms and egg turning status. The unique laminar airflow, coupled with high levels of insulation mean that temperature is extremely evenly distributed throughout the egg chamber, ensuring excellent hatch rates time after time. It comes with many sophisticated features which include room temperature alarms and a single button press to automatically level the shelves for easy tray access. An external humidity pump can also be added to provide fully automatic control of humidity.
Catalog No. NC1665287