Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ovalbumin-Biotin | 98.0% | 1 ML
SDP

Supplier:  Medchemexpress LLC HYNP0521ML

Encompass_Preferred

Ovalbumin-Biotin is a biotinylated biochemical reagent utilized in research to label or detect specific antigens, molecules, or cellular structures.

  • Purity: 98.0%
  • Appearance: Liquid
  • Color: Colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Store under recommended conditions in the Certificate of Analysis

Catalog No. 50-004-25776


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.