Learn More
Medchemexpress LLC Ovalbumin-Biotin | ≥98.0% | 100 UL
SDP
Supplier:  Medchemexpress LLC HYNP052100UL
Encompass_Preferred
Ovalbumin-Biotin is a biotinylated biochemical reagent. It is used to label or detect specific antigens, molecules, or cellular structures. This product is formulated in a solution containing 0.01 M TBS, pH 7.4 with 0.03% Proclin 300 and 50% Glycerol. This product is for research use only.
  • Used to label specific antigens
  • Used to detect specific antigens
  • Used to label specific molecules
  • Used to detect specific molecules
  • Used to label cellular structures
  • Used to detect cellular structures
  • Formulated in 0.01 M TBS, pH 7.4 with 0.03% Proclin 300 and 50% Glycerol
Catalog No. 50-004-26046