Learn More
Chemglass Life Sciences Vessel, Bio Reactor, 3L, Low Profile, Graduated, 100-440 GPI Thread CN, (6) Vertical GL-45 SN's
SDP
Supplier:  Chemglass Life Sciences CLS140603
Encompass_Preferred
Vessel has six vertical GL-45 threaded side necks and a 100mm threaded center neck.The 3 and 6 liter vessels are low profile to maximize incubator space.Supplied complete with (6) solid GL-45 caps and (6) pouring rings.
Catalog No. 50-121-4683