Learn More
Raybiotech Inc G-Series Ovine Cytokine Array 1 Kit.
Supplier:  Raybiotech Inc GSOCYT11
Encompass_Preferred
Semi-quantitative measurement of 10 ovine (sheep) cytokines. Suitable for all liquid sample types. Semi-Quantitative, Sandwich-based, Glass Slide Antibody Array Screen and compare protein expression levels with high specificity and sensitivity (pg/ml levels). The glass slide array allows for high throughput and low sample consumption; as little as 10 µl of original sample required per array. Proteins detected: Decorin, IFN-gamma, IL-17A, IL-21, IL-8, IP-10 (CXCL10), MIG (CXCL9), sFRP-3, TNF alpha, VEGF-A
Catalog No. 50-195-0478