Learn More
Raybiotech Inc Ovine IL-17A ELISA Kit
Supplier:  Raybiotech Inc ELOIL17A1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.45 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Ovine IL-17A.
Catalog No. 50-195-0169