Learn More
ACROBiosystems Recombinant Protein;Human OX40 / TNFRSF4 / CD134 Protein;HEK293;1MG;OX0,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Supplier:  ACROBiosystems OX0H52241MG
Encompass_Preferred
Recombinant Protein;Human OX40 / TNFRSF4 / CD134 Protein;1MG;Human OX40, His Tag (OX0-H5224) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # P43489-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40 Ligand, Fc Tag (Cat. No. OXL-H526x) at 5 μg/mL (100 μL/well) can bind Human OX40 (Cat. No. OX0-H5224) with a linear range of 0.128-2 μg/mL (QC tested).;FACS assay shows that recombinant Human OX40 (Cat. No. OX0-H5224) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 used is 0.3 μg/ml (Routinely tested).
Catalog No. 50-165-8526