Learn More
ACROBiosystems Recombinant Protein;Human OX40 / TNFRSF4 / CD134 Protein, Mouse IgG2a Fc Tag, low endotoxin;HEK293;100UG;OX0,TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L,OX-40
Supplier:  ACROBiosystems OX0H5252100UG
Encompass_Preferred
Recombinant Protein;Human OX40 / TNFRSF4 / CD134 Protein, Mouse IgG2a Fc Tag, low endotoxin;100UG;Human OX40, Mouse IgG2a Fc Tag, low endotoxin (OX0-H5252) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # P43489-1).;This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 47.1 kDa. The protein migrates as 58-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human OX40 Ligand, His Tag (Cat. No. OXL-H52Q8) at 2 μg/mL (100 μL/well) can bind Human OX40, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. OX0-H5252) with a linear range of 0.78-25 ng/mL (QC tested).;FACS assay shows that recombinant Human OX40, Mouse IgG2a Fc Tag (Cat. No. OX0-H5252) can bind to 293T cell overexpressing human OX40L. The concentration of OX40 is 0.01 μg/ml (Routinely tested).
Catalog No. 50-165-8003