Learn More
Medchemexpress LLC Oxaceprol | 33996-33-7 | 99.7% | 173.17 | 10 G
SDP
Supplier:  Medchemexpress LLC HY1749010G
Encompass_Preferred
Oxaceprol is an orally active derivative of L-proline that exhibits distinct anti-inflammatory activity. It is commonly used in research related to osteoarthritis and rheumatoid arthritis. It significantly reduces both the surface area and wet weight of cotton pellet granulomas in vivo.
  • Orally active derivative of L-proline
  • Possesses distinct anti-inflammatory activity
  • Reduces surface area and wet weight of cotton pellet granulomas
  • Inhibits secondary lesions in ears and tail
  • Does not inhibit prostaglandin synthesis in vitro
  • Markedly inhibits neutrophil infiltration into the joints of rats with adjuvant arthritis
  • Produced comparable inhibition of hyperalgesia to indomethacin
Catalog No. 50-003-86774