Learn More
Research Products International Corp Oxacillin Sodium Salt Monohydrate, 5 Grams
SDP
Supplier:  Research Products International Corp O210005.0
Encompass_Preferred
Oxacillin Sodium Salt Monohydrate is a significant compound playing a critical role in microbiology and pharmaceutical research. This specific formulation of oxacillin is instrumental in studying bacterial resistance mechanisms and antibiotic development. 
In microbiology, Oxacillin Sodium Salt Monohydrate is extensively employed to investigate bacterial susceptibility and resistance patterns. It is often used in laboratory settings to assess the effectiveness of antibiotics against various bacterial strains. This research aids in understanding the complex dynamics of microbial biology, including mechanisms of resistance, and contributes to the development of novel antibacterial strategies. 
Within pharmaceutical research, this compound serves as a reference standard for antibiotic formulation and efficacy testing, supporting the development of new antibiotics and strategies to combat antibiotic-resistant bacteria.
Catalog No. 50-997-950