Learn More
Apexbio Technology LLC Oxaliplatin, 61825-94-3, MFCD00866327, 50 mg
SDP
Supplier:  Apexbio Technology LLC A864850
Encompass_Preferred
MF: C8H14N2O4Pt, MW: 397.29, Solubility: 37.25mg/mL in DMSO, 3.94mg/mL in H2O with gentle warming. Oxaliplatin inhibits DNA synthesis, primarily by conforming DNA adducts. Oxaliplatin, a platinum compound, has a broad spectrum of anti-tumor activity and has demonstrated a lack of cross-resistance with other platinum compounds. Oxaliplatin induces primary and secondary DNA lesions that lead to cell apoptosis.
Catalog No. 50-101-1484