Learn More
Medchemexpress LLC Oxaliplatin | 61825-94-3 | 100.0% | 397.29 g/mol | C8H14N2O4Pt | 10 MG
SDP
Supplier:  Medchemexpress LLC HY17371R10MG
Encompass_Preferred
Oxaliplatin (Standard) is an analytical reference standard of the platinum-based chemotherapeutic agent oxaliplatin, supplied for research and analytical applications. It is intended for use as a calibration standard, quality-control material, and reference in assays assessing DNA crosslinking, DNA synthesis inhibition, and apoptosis.
  • High purity (99.97%).
  • Molecular formula C8H14N2O4Pt; molecular weight 397.29 g/mol.
  • Supplied in small package sizes suitable for analytical use.
  • Certificate of analysis and safety data sheet available.
  • Intended for research and analytical applications only.
  • Store and handle per COA and SDS; follow documented handling instructions.
Catalog No. 50-002-98717