Learn More
Medchemexpress LLC Oxaliplatin | 61825-94-3 | 99.99% | 397.29 | 200 MG
SDP
Supplier:  Medchemexpress LLC 200MGHY17371
Encompass_Preferred
Oxaliplatin is an analytical standard and DNA synthesis inhibitor. It induces DNA crosslinking damage, preventing DNA replication and transcription, and ultimately leading to apoptosis. This product is intended for various research and analytical applications, particularly in cancer research.
  • DNA synthesis inhibitor
  • Causes DNA crosslinking damage
  • Prevents DNA replication and transcription
  • Induces apoptosis
  • Suitable for cancer research
  • Analytical standard grade for reference assays
  • Used in qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
Catalog No. 50-003-65867