Learn More
Bioassay Systems EnzyChrom™ Oxaloacetate Assay Kit. For quantitative determination of oxaloacetate and evaluation of drug effects on its metabolism.
SDP
Supplier:  Bioassay Systems EOAA100
Encompass_Preferred
For quantitative determination of oxaloacetate and evaluation of drug effects on its metabolism. Key Features: Sensitive and accurate. Linear detection range: 7 to 400 µM oxaloacetate for colorimetric assays and 1 to 40 µM for fluorimetric assays. Method: OD570nm, or FL530/585nm. Samples: Plasma, serum, tissue and culture media. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: OD, FL: 4, 1 µM.
Catalog No. 50-489-284