Learn More
Medchemexpress LLC Oxaloacetate decarboxylase | 9024-98-0 | 1 KU
SDP
Supplier:  Medchemexpress LLC HYP28371KU
Encompass_Preferred
Oxaloacetate decarboxylase catalyzes the irreversible decarboxylation of oxaloacetate to pyruvate and COIC2. It is a member of the sodium ion transport decarboxylase (NaT-DC) enzyme family and converts the chemical energy of the decarboxylation reaction into an electrochemical gradient of Na+ ions across the membrane.
  • Catalyzes irreversible decarboxylation of oxaloacetate to pyruvate and COIC2
  • Member of the sodium ion transport decarboxylase (NaT-DC) enzyme family
  • Converts chemical energy of decarboxylation reaction into an electrochemical gradient of Na+ ions
Catalog No. 50-003-97405