Learn More
Medchemexpress LLC Oxaloacetate decarboxylase | 9024-98-0 | 5 KU
SDP
Supplier:  Medchemexpress LLC HYP28375KU
Encompass_Preferred
Oxaloacetate decarboxylase catalyzes the irreversible decarboxylation of oxaloacetate to pyruvate and COIC2. This enzyme belongs to the sodium ion transport decarboxylase (NaT-DC) enzyme family and converts the chemical energy from the decarboxylation reaction into an electrochemical gradient of Na+ ions across the membrane.
  • Catalyzes irreversible decarboxylation of oxaloacetate to pyruvate and COIC2.
  • Member of the sodium ion transport decarboxylase (NaT-DC) enzyme family.
  • Converts chemical energy of decarboxylation into an electrochemical gradient of Na+ ions.
  • Available in various sizes: 500 U, 1 KU, 5 KU.
  • For research use only.
Catalog No. 50-004-00307