Learn More
Medchemexpress LLC Oxaloacetate decarboxylase | 9024-98-0 | 500 U
SDP
Supplier:  Medchemexpress LLC HYP2837500U
Encompass_Preferred
Oxaloacetate decarboxylase catalyzes the irreversible decarboxylation of oxaloacetate to pyruvate and CO₂. It is a member of the sodium ion transport decarboxylase (NaT-DC) enzyme family.
  • Converts the chemical energy of the decarboxylation reaction into an electrochemical gradient of Na⁺ ions across the membrane.
  • For research use only.
Catalog No. 50-003-96789