Learn More
Medchemexpress LLC Oxalomalic acid trisodium | 89304-26-7 | MFCD00056721 | 95.0% | C6H3Na3O8 | 10MG

Supplier: Medchemexpress LLC HY13152110MG
Oxalomalic acid trisodium is the trisodium salt of oxalomalic acid (oxalomalate) used in biochemical research as an inhibitor of aconitase and NADP-dependent isocitrate dehydrogenase (IDH). Supplied as a light yellow to yellow solid, it is provided with specified purity and storage recommendations for research applications.
- Inhibits aconitase and NADP-dependent isocitrate dehydrogenase.
- Supplied as a light yellow to yellow solid.
- Purity 95.0%.
- Molecular formula C6H3Na3O8; molecular weight 272.05.
- Recommended storage: sealed, away from moisture; in solvent: -80°C (6 months), -20°C (1 month).
- Available in small pack sizes suitable for research use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.