Learn More
Abcam O x amflatin, HDAC inhibitor, 1MG
Supplier:  Abcam AB1445071MG
Encompass_Preferred
MW 342.4 Da, Purity >95%. Potent mammalian HDAC inhibitor (IC₅₀ = 15.7 nM). Induces an elongated cell shape morphology with filamentous protrusions. Shows antiproliferative effects. Shows antitumor effects in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5661