Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxamic acid sodium | 565-73-1 | 99.7% | 111.03 | 5 G
SDP

Supplier:  Medchemexpress LLC HYW013032A5G

Encompass_Preferred

Oxamic acid sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor. This compound demonstrates anti-tumor and anti-proliferative activities against cancer cells, and is capable of inducing apoptosis, making it suitable for research applications.

  • Inhibits lactate dehydrogenase-A (LDH-A).
  • Exhibits anti-tumor activity.
  • Demonstrates anti-proliferative activity against cancer cells.
  • Induces apoptosis.
  • Appears as a white to off-white solid.
  • Soluble in water and DMSO with specific conditions.
  • Store at 4°C, sealed and away from moisture for optimal stability.
  • When in solvent, store at -80°C for up to 6 months or -20°C for 1 month.

Catalog No. 50-004-54094


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.