Learn More
Apexbio Technology LLC Oxaprozin 21256-18-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18045.1
Encompass_Preferred
Oxaprozin (CAS 21256-18-8) is a non-steroidal anti-inflammatory agent that functions primarily through inhibition of cyclooxygenase (COX) enzymes leading to reduced synthesis of pro-inflammatory prostaglandins This mechanism underlies its capacity to attenuate inflammation swelling and joint pain in conditions such as osteoarthritis and rheumatoid arthritis Oxaprozin is utilized in biomedical research to model and investigate inflammatory processes evaluate analgesic pathways and explore COX-mediated signaling in disease pathology
Catalog No. 50-000-91796