Learn More
Medchemexpress LLC Oxaprozin-d4 | 21256-18-8 | MFCD00215977 | 99.1% | 297.34 g·mol^-1 | C18H11D4NO3 | 10 MG

Supplier: Medchemexpress LLC HYB0808S10MG
Oxaprozin-d4 is the deuterium-labeled isotopologue of oxaprozin (d4), a non-steroidal anti-inflammatory drug and cyclooxygenase (COX) inhibitor supplied as a solid research reagent. It is intended for research use only and is commonly used as an internal standard or tracer in analytical and pharmacokinetic studies.
- Deuterium-labeled oxaprozin for tracer and quantitative analysis.
- Supplied as a 10 mg solid suitable for analytical workflows.
- High purity (approximately 99.1%) to minimize analytical interference.
- Chemical formula C18H11D4NO3 and molecular weight 297.34 g·mol^-1.
- Stable when stored as recommended: powder at -20°C for long-term storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.