Learn More
Medchemexpress LLC HY-123205 5mg Medchemexpress, Oxatomide CAS:60607-34-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1232055MG
Encompass_Preferred
Medchemexpress, HY-123205 5mg Oxatomide CAS:60607-34-3 Oxatomide is a potent and orally active dual H1-histamine receptor and P2X7 receptor antagonist with antihistamine and anti-allergic activity. Oxatomide almost completely blocks the ATP-induced current in human P2X7 receptors (IC50 of 0.95 μM). Oxatomide inhibits ATP-induced Ca2+ influx with an IC50 value of 0.43 μM and also inhibits serotonin. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1008