Learn More
Cayman Chemical OxcarbazepIn-d4 1mg
SDP
Supplier:  Cayman Chemical 299951 MG
Encompass_Preferred
An internal standard for the quantification of oxcarbazepine by GC- or LC-MS
Catalog No. 50-298-9837