Learn More
Medchemexpress LLC Oxcarbazepine | 28721-07-5 | 99.9% | C15H12N2O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYB01145MG
Encompass_Preferred
Oxcarbazepine is a sodium channel blocker that significantly inhibits glioblastoma cell growth and induces apoptosis or G2/M arrest in glioblastoma cell lines. It exhibits anti-cancer and anticonvulsant effects. In vitro, it significantly inhibits glioblastoma cell growth with IC50 values of 12.35 μg/mL for U87 cell lines and 9.45 μg/mL for T98 cell lines.
  • Sodium channel blocker
  • Inhibits glioblastoma cell growth
  • Induces apoptosis or G2/M arrest in glioblastoma cell lines
  • Exhibits anti-cancer effects
  • Exhibits anticonvulsant effects
  • Protects against generalized tonic-clonic seizures
Catalog No. 50-004-33348