Learn More
Apexbio Technology LLC Oxeladin Citrate 52432-72-1 5g
SDP
Supplier:  Apexbio Technology LLC A84935000
Encompass_Preferred
Oxeladin citrate (CAS 52432-72-1) is a small-molecule agonist targeting sigma-1 receptors It is designed to selectively activate sigma-1 receptors thereby modulating neural signaling pathways involved in cough reflex regulation neuroprotection pain perception and respiratory control Oxeladin citrate exerts its biological activity primarily through sigma-1 receptor agonism It exhibits an affinity (Ki) of approximately 25 nM for the sigma-1 receptor Based on these pharmacological properties Oxeladin citrate holds research potential in investigating cough suppression respiratory function and pathophysiological conditions associated with central nervous system pulmonary dysfunction and cerebrovascular disorders
Catalog No. 50-000-95639