Learn More
Cayman Chemical Oxfendazole 50mg
SDP
Supplier:  Cayman Chemical 2974250 MG
Encompass_Preferred
An anthelmintic; curative in pig models of H. rubidus or A. suum infection at 3 and 6 mg/kg, respectively; is lethal to H. contortus and T. circumcincta when administered to infected cattle (LD50s = 4.28 and 3.61 mg/kg of host weight, respectively)
Catalog No. 50-298-8370