Learn More
Apexbio Technology LLC Oxfendazole 53716-50-0 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B19985.1
Encompass_Preferred
Oxfendazole (CAS 53716-50-0) is a small-molecule inhibitor targeting parasite tubulin It is designed to inhibit microtubule polymerization thereby disrupting microtubule function and promoting parasite elimination Oxfendazole exerts its biological activity primarily through binding to tubulin and inhibiting parasite microtubule polymerization In in vitro studies Oxfendazole demonstrates antiparasitic and antitumor activity with IC50 values typically ranging from low micromolar to submicromolar levels depending on the specific cell lines and experimental settings Based on these pharmacological properties Oxfendazole holds research potential in the investigation of antiparasitic activity tumor cell growth inhibition and mechanistic studies of microtubule destabilization
Catalog No. 50-000-93294