Learn More
Medchemexpress LLC Oxfendazole | 53716-50-0 | >98.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0291100MG
Encompass_Preferred
Oxfendazole is a sulfoxide form of fenbendazole, which is effective when taken orally. This compound fights parasites and has demonstrated tumor-promoting activity. It is intended for research use only.
  • Sulfoxide form of fenbendazole
  • Effective when taken orally
  • Fights parasites
  • Exhibits tumor-promoting activity
  • Appears as a white to off-white solid
  • Soluble in DMSO at 31.25 mg/mL
  • Soluble in H2O at 0.67 mg/mL
  • Involved in clinical trials for conditions such as trichuriasis and brain cysticercosis
Catalog No. 50-004-06025