Learn More
Medchemexpress LLC Oxfendazole | 53716-50-0 | 98.8% | 5 G
SDP
Supplier:  Medchemexpress LLC HYB02915G
Encompass_Preferred
Oxfendazole is a sulfoxide form of fenbendazole that is effective when taken orally. It fights parasites and exhibits tumor-promoting activity.
  • Effective orally.
  • Antiparasitic agent.
  • Exhibits tumor-promoting activity.
  • Intended for research use only.
Catalog No. 50-004-07020