Learn More
Medchemexpress LLC Oxfendazole | 53716-50-0 | 98.8% | 500 MG
SDP
Supplier:  Medchemexpress LLC HYB0291500MG
Encompass_Preferred
Oxfendazole is a sulfoxide form of fenbendazole that is effective when taken orally. It fights parasites and has tumor-promoting activity. This product is for research use only.
  • Sulfoxide form of fenbendazole
  • Effective when taken orally
  • Fights parasites
  • Promotes tumor activity
  • For research use only
  • Effective against gastrointestinal nematodes in pigs
Catalog No. 50-004-06474