Learn More
Medchemexpress LLC Oxfendazole | 53716-50-0 | MFCD00801063 | 98.8% | 1 G

Supplier: Medchemexpress LLC HYB02911G
Oxfendazole is a sulfoxide form of fenbendazole that is effective when taken orally. It fights parasites and has tumor-promoting activity.
- Appearance: White to off-white solid
- Purity: 98.8% (LCMS)
- Molecular Weight: 315.35
- Molecular Formula: C15H13N3O3S
- Consistent with structure by 1H NMR and LCMS
- Intended for laboratory chemicals and substance manufacturing
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.